Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gjb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008125.3 | NP_032151.1 | 2 | 896 | ||
GenBank mRNA | AK133942.1 | - | 2 | 811 | ||
AK135055.1 | - | 2 | 897 | |||
AK146485.1 | - | 2 | 897 | |||
AK157920.1 | - | 2 | 897 | |||
AK160905.1 | - | 2 | 897 | |||
AK178984.1 | - | 1 | 148 | |||
AK189228.1 | - | 1 | 148 | |||
BC013634.1 | - | 2 | 871 | 4194757 | ||
BC051437.1 | - | 2 | 841 | 6311829 |