Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMMR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142556.1 | NP_001136028.1 | 14 | 1829 | ||
NM_001142557.1 | NP_001136029.1 | 11 | 1602 | |||
NM_012484.2 | NP_036616.2 | 14 | 1826 | |||
NM_012485.2 | NP_036617.2 | 13 | 1781 | |||
GenBank mRNA | AF032862.1 | - | 14 | 1680 | ||
AK302975.1 | - | 15 | 1636 | |||
AK303616.1 | - | 11 | 1480 | |||
AK313603.1 | - | 14 | 1747 | |||
AY623426.1 | - | 14 | 1652 | |||
BC033568.1 | - | 4 | 515 | |||
BC057235.1 | - | 13 | 1751 | |||
BC070281.1 | - | 13 | 1657 | |||
BC108904.1 | - | 14 | 1681 | 40007903 | ||
U29343.1 | - | 14 | 1707 |