Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ostm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172416.3 | NP_766004.1 | 5 | 1009 | ||
GenBank mRNA | AF533890.1 | - | 5 | 1008 | ||
AK004546.1 | - | 5 | 986 | |||
AK044897.1 | - | 5 | 1008 | |||
AK050513.1 | - | 5 | 983 | |||
AK053664.1 | - | 4 | 2648 | |||
BC057635.1 | - | 5 | 947 | 4910872 |