Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gli1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010296.2 | NP_034426.2 | 13 | 2476 | ||
GenBank mRNA | AB025922.1 | - | 13 | 2486 | ||
AF026305.1 | - | 13 | 2473 | |||
BC131650.1 | - | 12 | 2454 | 40110429 |