Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gli2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081125.1 | NP_001074594.1 | 9, 10 | 1659 | ||
GenBank mRNA | BC082604.1 | - | 9, 10 | 1600 | 30672479 | |
BC085190.1 | - | 9, 10 | 1575 | 30694633 | ||
X99104.1 | - | 9, 10 | 1419 |