Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gna11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010301.3 | NP_034431.1 | 5 | 764 | ||
GenBank mRNA | AK139289.1 | - | 1 | 603 | ||
AK143685.1 | - | 5 | 765 | |||
BC011169.1 | - | 5 | 763 | 4206878 | ||
M55411.1 | - | 5 | 701 | |||
M57617.1 | - | 1 | 15 |
Gene Symbol: Gnaz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | L17074.1 | - | 1 | 16 |