Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gna11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010301.3 | NP_034431.1 | 2 | 354 | ||
GenBank mRNA | AK143685.1 | - | 2 | 355 | ||
BC011169.1 | - | 2 | 353 | 4206878 | ||
M55411.1 | - | 2 | 291 |