Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202233.1 | NP_001189162.1 | 3 | 942 | ||
NM_001202234.1 | NP_001189163.1 | 3 | 1869 | |||
NM_002135.4 | NP_002126.2 | 3 | 1031 | |||
NM_173157.2 | NP_775180.1 | 2 | 881 | |||
XM_005268822.3 | XP_005268879.1 | 3 | 1011 | |||
XM_005268824.3 | XP_005268881.1 | 2 | 1201 | |||
XM_006719363.1 | XP_006719426.1 | 3 | 937 | |||
XM_006719364.3 | XP_006719427.1 | 4 | 1008 | |||
XM_017019247.1 | XP_016874736.1 | 2 | 813 | |||
GenBank mRNA | AB307717.1 | - | 1 | 712 | ||
AK075491.1 | - | 2 | 824 | |||
AK131566.1 | - | 3 | 1871 | |||
AK297526.1 | - | 3 | 943 | |||
AK314437.1 | - | 2 | 835 | |||
BC016147.1 | - | 3 | 976 | 3921259 | ||
BT007144.1 | - | 1 | 712 | |||
CR456704.1 | - | 1 | 712 | |||
D49728.1 | - | 2 | 821 | |||
D85245.1 | - | 2 | 902 | |||
HQ692855.1 | - | 1 | 712 | |||
L13740.1 | - | 2 | 822 |