Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnai2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008138.4 | NP_032164.2 | 8 | 1060 | ||
GenBank mRNA | AK157998.1 | - | 8 | 1064 | ||
AK159222.1 | - | 8 | 1064 | |||
AK160186.1 | - | 8 | 1057 | |||
AK167388.1 | - | 8 | 1062 | |||
AK179809.1 | - | 2 | 209 | |||
AK212898.1 | - | 2 | 113 | |||
BC006695.1 | - | 1 | 31 | |||
BC037130.1 | - | 4 | 384 | |||
BC065159.1 | - | 8 | 1047 | 5717305 | ||
M13963.1 | - | 8 | 1032 |