Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnai3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010306.3 | NP_034436.1 | 4 | 577 | ||
GenBank mRNA | AK004566.1 | - | 4 | 571 | ||
AK077490.1 | - | 4 | 539 | |||
AK133749.1 | - | 4 | 518 | |||
AK167438.1 | - | 4 | 531 | |||
AK168578.1 | - | 4 | 515 | |||
AK169067.1 | - | 4 | 522 | |||
AK169285.1 | - | 4 | 552 | |||
BC003462.1 | - | 4 | 503 | |||
BC020009.1 | - | 4 | 549 | |||
BC041107.1 | - | 4 | 488 | 2648164 |