Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnai3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010306.3 | NP_034436.1 | 5 | 621 | ||
GenBank mRNA | AK004566.1 | - | 5 | 615 | ||
AK077490.1 | - | 5 | 583 | |||
AK133749.1 | - | 5 | 562 | |||
AK167438.1 | - | 5 | 575 | |||
AK168578.1 | - | 5 | 559 | |||
AK169067.1 | - | 5 | 566 | |||
AK169285.1 | - | 5 | 596 | |||
BC003462.1 | - | 5 | 547 | |||
BC020009.1 | - | 5 | 593 | |||
BC041107.1 | - | 5 | 532 | 2648164 |