Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnai3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010306.3 | NP_034436.1 | 7 | 965 | ||
GenBank mRNA | AK004566.1 | - | 7 | 959 | ||
AK077490.1 | - | 7 | 927 | |||
AK133749.1 | - | 7 | 906 | |||
AK167438.1 | - | 7 | 919 | |||
AK168578.1 | - | 7 | 903 | |||
AK169067.1 | - | 7 | 910 | |||
AK169285.1 | - | 7 | 940 | |||
BC020009.1 | - | 7 | 937 | |||
BC041107.1 | - | 7 | 876 | 2648164 | ||
U38502.1 | - | 1 | 79 |