Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013531.4 | NP_038559.2 | 10 | 1080 | ||
GenBank mRNA | AF277893.1 | - | 9 | 944 | ||
AF501886.1 | - | 9 | 944 | |||
AK036816.1 | - | 10 | 1082 | |||
AK098139.1 | - | 10 | 1013 | |||
AK152227.1 | - | 10 | 1071 | |||
AK155309.1 | - | 10 | 1139 | |||
AK158965.1 | - | 11 | 1310 | |||
AK167416.1 | - | 10 | 1113 | |||
AK168302.1 | - | 10 | 1036 | |||
AK169752.1 | - | 9 | 1157 | |||
AK172147.1 | - | 11 | 1213 | |||
BC028753.1 | - | 10 | 1051 | 1348753 | ||
M63658.1 | - | 9 | 984 | |||
M87286.1 | - | 9 | 984 | |||
S86124.1 | - | 9 | 984 |