Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013531.4 | NP_038559.2 | 9 | 930 | ||
GenBank mRNA | AF277893.1 | - | 8 | 794 | ||
AF501886.1 | - | 8 | 794 | |||
AK036816.1 | - | 9 | 932 | |||
AK098139.1 | - | 9 | 863 | |||
AK152227.1 | - | 9 | 921 | |||
AK155309.1 | - | 9 | 989 | |||
AK158965.1 | - | 10 | 1160 | |||
AK167416.1 | - | 9 | 963 | |||
AK168302.1 | - | 9 | 886 | |||
AK169752.1 | - | 8 | 1007 | |||
AK172147.1 | - | 10 | 1063 | |||
BC028753.1 | - | 9 | 901 | 1348753 | ||
M63658.1 | - | 8 | 834 | |||
M87286.1 | - | 8 | 834 | |||
S86124.1 | - | 8 | 834 |