Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013531.4 | NP_038559.2 | 5 | 390 | ||
GenBank mRNA | AF277893.1 | - | 4 | 254 | ||
AF501886.1 | - | 4 | 254 | |||
AK036816.1 | - | 5 | 392 | |||
AK098139.1 | - | 5 | 323 | |||
AK152227.1 | - | 5 | 381 | |||
AK155309.1 | - | 5 | 449 | |||
AK158965.1 | - | 6 | 620 | |||
AK165084.1 | - | 4 | 3437 | |||
AK167416.1 | - | 5 | 423 | |||
AK168302.1 | - | 5 | 346 | |||
AK169752.1 | - | 5 | 381 | |||
AK172147.1 | - | 6 | 523 | |||
AK187079.1 | - | 2 | 155 | |||
AK198808.1 | - | 2 | 154 | |||
BC028753.1 | - | 5 | 361 | 1348753 | ||
M87286.1 | - | 4 | 294 | |||
S86124.1 | - | 4 | 294 |