Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnpat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010322.3 | NP_034452.3 | 7 | 1019 | ||
GenBank mRNA | AF110769.1 | - | 7 | 983 | ||
AJ132012.1 | - | 7 | 814 | |||
AK004630.1 | - | 7 | 993 | |||
AK004771.1 | - | 7 | 999 | |||
AK010896.1 | - | 7 | 993 | |||
AK150810.1 | - | 7 | 977 | |||
AK166997.1 | - | 7 | 991 | |||
AK178085.1 | - | 2 | 124 | |||
AK181282.1 | - | 2 | 125 | |||
BC003225.1 | - | 7 | 961 | |||
BC025972.1 | - | 7 | 976 | 4020505 |