Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnrhr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010323.1 | NP_034453.1 | 3 | 917 | ||
GenBank mRNA | L01119.1 | - | 3 | 917 | ||
L28756.1 | - | 736 | ||||
L28758.1 | - | 3 | 872 | |||
M93108.1 | - | 3 | 914 |
Set Membership: |
G-protein coupled receptor |