Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Got2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010325.2 | NP_034455.1 | 2 | 266 | ||
GenBank mRNA | AK098166.1 | - | 2 | 290 | ||
AK136556.1 | - | 2 | 303 | |||
AK147953.1 | - | 2 | 306 | |||
AK149886.1 | - | 2 | 303 | |||
AK149926.1 | - | 2 | 303 | |||
AK150194.1 | - | 1 | 22 | |||
AK152921.1 | - | 2 | 303 | |||
AK155075.1 | - | 2 | 297 | |||
BC089015.1 | - | 2 | 292 | 30606379 | ||
BC089341.1 | - | 2 | 304 | 6832287 | ||
J02622.1 | - | 2 | 266 | |||
U82470.1 | - | 2 | 234 |