Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lilrb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291894.1 | NP_001278823.1 | 827 | |||
NM_013532.3 | NP_038560.1 | 822 | ||||
GenBank mRNA | AK170368.1 | - | 708 | |||
BC006591.1 | - | 723 | 3585404 | |||
BC099532.1 | - | 714 | 30749706 | |||
U05264.1 | - | 723 |
Gene Symbol: Gp49a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291892.1 | NP_001278821.1 | 5, 6 | 815 | ||
NM_001291893.1 | NP_001278822.1 | 4, 5 | 525 | |||
NM_008147.2 | NP_032173.1 | 5, 6 | 810 | |||
GenBank mRNA | AK089366.1 | - | 4, 5 | 362 | ||
AK167751.1 | - | 5, 6 | 731 | |||
AK171573.1 | - | 5, 6 | 759 | |||
BC065784.1 | - | 4, 5 | 479 | |||
M65027.1 | - | 5, 6 | 743 |