Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008154.3 | NP_032180.1 | 2 | 903 | ||
GenBank mRNA | AK140302.1 | - | 2 | 874 | ||
AK140464.1 | - | 2 | 857 | |||
AK158917.1 | - | 2 | 866 | |||
BC138248.1 | - | 1 | 1225 | 8861267 | ||
BC138250.1 | - | 1 | 1225 | 8861269 | ||
D21062.1 | - | 1 | 1278 |
Set Membership: |
G-protein coupled receptor |