Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008155.4 | NP_032181.2 | 12 | 1098 | ||
GenBank mRNA | AK076424.1 | - | 10 | 754 | ||
AK133827.1 | - | 12 | 1020 | |||
AK134890.1 | - | 12 | 1032 | |||
AK137805.1 | - | 12 | 1026 | |||
AK147124.1 | - | 12 | 1061 | |||
AK150341.1 | - | 12 | 1029 | |||
AK152508.1 | - | 7 | 402 | |||
AK159838.1 | - | 12 | 1002 | |||
AK169681.1 | - | 12 | 1027 | |||
AK197318.1 | - | 2 | 82 | |||
BC086640.1 | - | 12 | 1045 | 6832250 | ||
BC088995.1 | - | 12 | 1005 | 6403790 | ||
L09104.1 | - | 3 | 2237 | |||
M14220.1 | - | 12 | 1027 | |||
U89408.1 | - | 5 | 273 |