Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpm6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177955.1 | NP_001171426.1 | 8 | 1404 | ||
NM_001177956.1 | NP_001171427.1 | 8 | 2965 | |||
NM_001177957.1 | NP_001171428.1 | 9 | 1495 | |||
NM_001177958.1 | NP_001171429.1 | 7 | 1284 | |||
NM_001177959.1 | NP_001171430.1 | 7 | 2845 | |||
NM_001177960.1 | NP_001171431.1 | 8 | 1375 | |||
NM_001177961.1 | NP_001171432.1 | 7 | 2510 | |||
NM_001177962.1 | NP_001171433.1 | 8 | 1040 | |||
NM_023122.3 | NP_075611.1 | 7 | 949 | |||
GenBank mRNA | AF254869.1 | - | 7 | 1145 | ||
AF254870.1 | - | 8 | 1265 | |||
AF254871.1 | - | 7 | 920 | |||
AF254872.1 | - | 8 | 1236 | |||
AF254873.1 | - | 7 | 2705 | |||
AF254874.1 | - | 9 | 1356 | |||
AF254875.1 | - | 8 | 2825 | |||
AF254876.1 | - | 8 | 1011 | |||
AF254877.1 | - | 7 | 2480 | |||
AK016567.1 | - | 7 | 874 | |||
AK140714.1 | - | 7 | 964 | |||
S65737.1 | - | 7 | 941 |