Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030677.2 | NP_109602.2 | 1 | 312 | ||
GenBank mRNA | AK131940.1 | - | 1 | 189 | ||
BC010823.1 | - | 1 | 182 | 4218623 | ||
BC034335.1 | - | 1 | 159 | 5038341 | ||
BC039658.1 | - | 1 | 257 | 4987478 | ||
BC054848.1 | - | 1 | 135 | 6529267 | ||
BC066825.1 | - | 1 | 256 | |||
CF612186.1 | - | 1 | 340 |
Gene Symbol: Gpx2-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BG084322.2 | - | 1 | 255 |