Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008161.3 | NP_032187.2 | 2, 3 | 349 | ||
GenBank mRNA | AK002219.1 | - | 2, 3 | 278 | ||
AK002262.1 | - | 2, 3 | 278 | |||
AK004942.1 | - | 2, 3 | 278 | |||
AK146760.1 | - | 2, 3 | 278 | |||
BC003339.1 | - | 2, 3 | 261 | 3257333 | ||
BC037027.1 | - | 2, 3 | 296 | 5370640 | ||
BC037134.1 | - | 2, 3 | 275 | |||
BC049235.1 | - | 2, 3 | 265 | 4221025 | ||
BC061950.1 | - | 2, 3 | 253 | 6528998 | ||
BY061328.1 | - | 2, 3 | 350 | |||
CB601893.1 | - | 2, 3 | 281 | |||
U13705.1 | - | 2, 3 | 253 |