Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008163.3 | NP_032189.1 | 6 | 888 | ||
GenBank mRNA | AK077750.1 | - | 5 | 708 | ||
AK079087.1 | - | 5 | 716 | |||
AK148148.1 | - | 6 | 853 | |||
AK153557.1 | - | 5 | 821 | |||
AK155800.1 | - | 6 | 843 | |||
AK166142.1 | - | 6 | 842 | |||
AK170845.1 | - | 5 | 679 | |||
AK172002.1 | - | 6 | 864 | |||
AK192819.1 | - | 1 | 78 | |||
AK202005.1 | - | 1 | 78 | |||
BC052377.1 | - | 6 | 888 | 5696248 | ||
BC085254.1 | - | 6 | 724 | 5343274 | ||
D85748.1 | - | 4 | 512 | |||
U07617.1 | - | 5 | 637 |