Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008163.3 | NP_032189.1 | 5 | 749 | ||
GenBank mRNA | AK077750.1 | - | 4 | 569 | ||
AK079087.1 | - | 4 | 577 | |||
AK148148.1 | - | 5 | 714 | |||
AK153557.1 | - | 4 | 682 | |||
AK155800.1 | - | 5 | 704 | |||
AK166142.1 | - | 5 | 703 | |||
AK170845.1 | - | 4 | 540 | |||
AK172002.1 | - | 5 | 725 | |||
AK181485.1 | - | 1 | 42 | |||
AK212651.1 | - | 1 | 74 | |||
AK216513.1 | - | 1 | 74 | |||
AK217799.1 | - | 1 | 74 | |||
BC052377.1 | - | 5 | 749 | 5696248 | ||
BC085254.1 | - | 5 | 585 | 5343274 | ||
D85748.1 | - | 3 | 373 | |||
U07617.1 | - | 4 | 498 |