Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010346.2 | NP_034476.1 | 5 | 956 | ||
GenBank mRNA | AK057640.1 | - | 5 | 951 | ||
AK145996.1 | - | 5 | 772 | |||
AK168438.1 | - | 5 | 740 | |||
AK220277.1 | - | 5 | 722 | |||
BC003295.1 | - | 5 | 699 | 3583896 | ||
BC081547.1 | - | 5 | 742 |