Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aes | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276288.1 | NP_001263217.1 | 4 | 419 | ||
NM_010347.4 | NP_034477.1 | 4 | 419 | |||
GenBank mRNA | AK131845.1 | - | 4 | 296 | ||
AK131994.1 | - | 4 | 367 | |||
AK149015.1 | - | 1 | 1087 | |||
AK158714.1 | - | 4 | 304 | |||
BC132217.1 | - | 4 | 307 | 40130494 | ||
BC132219.1 | - | 4 | 307 | 40130496 | ||
BC145299.1 | - | 4 | 307 | 9053853 | ||
BE685863.1 | - | 4 | 211 | |||
BY713872.1 | - | 4 | 420 | |||
L12140.1 | - | 4 | 260 | |||
X73359.1 | - | 3 | 370 | |||
X73361.1 | - | 4 | 319 |