Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gria1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113325.2 | NP_001106796.1 | 10 | 1788 | ||
NM_001252403.1 | NP_001239332.1 | 10 | 1415 | |||
NM_008165.4 | NP_032191.2 | 10 | 1788 | |||
GenBank mRNA | AB102773.1 | - | 10 | 1372 | ||
AB102774.1 | - | 10 | 1372 | |||
AK049958.1 | - | 10 | 1598 | |||
AK135107.1 | - | 10 | 1416 | |||
BC056397.1 | - | 10 | 1450 | 6842391 | ||
BC060702.1 | - | 10 | 1371 | |||
X57497.1 | - | 10 | 1635 |