Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gria2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039195.1 | NP_001034284.1 | 8 | 1504 | ||
NM_001083806.1 | NP_001077275.1 | 8 | 1504 | |||
NM_013540.2 | NP_038568.2 | 8 | 1504 | |||
GenBank mRNA | AB109210.1 | - | 8 | 1097 | ||
AB109211.1 | - | 8 | 1097 | |||
AB111957.1 | - | 8 | 1097 | |||
AK014389.1 | - | 8 | 1549 | |||
AK043490.1 | - | 8 | 1477 | |||
AK044574.1 | - | 8 | 1436 | |||
AK046861.1 | - | 8 | 1504 | |||
AK086614.1 | - | 8 | 1438 | |||
AK134572.1 | - | 8 | 1434 | |||
AK134647.1 | - | 7 | 950 | |||
AK134772.1 | - | 7 | 1075 | |||
AK141525.1 | - | 8 | 1480 | |||
AK163941.1 | - | 8 | 1497 | |||
BC048248.1 | - | 8 | 1363 | 6494013 | ||
BC066193.1 | - | 8 | 1292 | |||
X57498.1 | - | 8 | 1299 |