Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008167.2 | NP_032193.1 | 16 | 2633 | ||
GenBank mRNA | AK032669.1 | - | 4 | 457 | ||
BC139823.1 | - | 16 | 2637 | 40141838 | ||
D13266.1 | - | 16 | 2633 |