Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grik1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010348.3 | NP_034478.1 | 2 | 753 | ||
NM_146072.4 | NP_666184.2 | 2 | 753 | |||
GenBank mRNA | AK042635.1 | - | 2 | 754 | ||
AK043542.1 | - | 2 | 733 | |||
AK043737.1 | - | 2 | 717 | |||
AK044250.1 | - | 2 | 712 | |||
AK044420.1 | - | 2 | 709 | |||
AK046122.1 | - | 2 | 760 | |||
AK048634.1 | - | 2 | 724 | |||
AK133667.1 | - | 2 | 739 | |||
BC031822.1 | - | 1 | 175 | 4501004 | ||
BC049275.1 | - | 1 | 175 | 4501004 | ||
X66118.1 | - | 2 | 739 |