Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grik3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081097.2 | NP_001074566.1 | 3 | 448 | ||
GenBank mRNA | BC118004.1 | - | 3 | 443 |