Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdia3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007952.2 | NP_031978.2 | 5 | 672 | ||
GenBank mRNA | AK088721.1 | - | 5 | 632 | ||
AK132459.1 | - | 5 | 637 | |||
AK133799.1 | - | 5 | 631 | |||
AK167807.1 | - | 5 | 625 | |||
AK169611.1 | - | 5 | 626 | |||
BC003285.1 | - | 5 | 591 | 3582199 | ||
BC033439.1 | - | 5 | 566 | 4016310 | ||
BQ714617.1 | - | 672 | ||||
M73329.1 | - | 5 | 611 |