Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002136.3 | NP_002127.1 | 10 | 1721 | ||
NM_031157.3 | NP_112420.1 | 11 | 1877 | |||
XM_017019251.1 | XP_016874740.1 | 9 | 1640 | |||
GenBank mRNA | AK129814.1 | - | 10 | 1663 | ||
BC002355.2 | - | 10 | 1642 | 2821751 | ||
BC009600.1 | - | 10 | 1664 | 4134638 | ||
BC012158.1 | - | 10 | 1624 | 4564655 | ||
BC033714.1 | - | 10 | 1653 | 4420039 | ||
BC035253.1 | - | 6 | 1080 | |||
BC052296.1 | - | 10 | 1633 | 6297642 | ||
BC074502.1 | - | 10 | 1646 | 3455742 | ||
BC103707.1 | - | 9 | 1472 | 6154800 | ||
X06747.1 | - | 10 | 1687 |
Gene Symbol: XKRX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011530954.2 | XP_011529256.1 | 4 | 4084 |