Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsta3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077353.2 | NP_001070821.1 | 8 | 807 | ||
NM_001288617.1 | NP_001275546.1 | 8 | 852 | |||
NM_010356.4 | NP_034486.2 | 7 | 712 | |||
GenBank mRNA | AK002478.1 | - | 6 | 660 | ||
AK014076.1 | - | 7 | 685 | |||
AK149543.1 | - | 7 | 677 | |||
BC147272.1 | - | 7 | 675 | 9056327 | ||
BC147273.1 | - | 7 | 675 | 9056339 | ||
M73483.1 | - | 6 | 643 | |||
X65021.1 | - | 6 | 628 |