Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008183.3 | NP_032209.1 | 4 | 234 | ||
GenBank mRNA | AK002845.1 | - | 4 | 251 | ||
BC019426.1 | - | 4 | 204 | |||
BC037068.1 | - | 4 | 258 | 4990201 | ||
J04696.1 | - | 4 | 195 |