Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252555.1 | NP_001239484.1 | 3, 4 | 487 | ||
NM_010363.4 | NP_034493.1 | 2, 3 | 184 | |||
GenBank mRNA | AB041613.1 | - | 3, 4 | 487 | ||
AF093418.1 | - | 2, 3 | 133 | |||
AK002398.1 | - | 2, 3 | 141 | |||
AK012796.1 | - | 2, 3 | 184 | |||
AK075927.1 | - | 2, 3 | 138 | |||
BC031777.1 | - | 2, 3 | 173 | 4168439 | ||
BY072664.1 | - | 2, 3 | 185 |