Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2h4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF054823.1 | - | 10 | 1061 | ||
AK079456.1 | - | 10 | 1090 | |||
AK088296.1 | - | 10 | 1087 | |||
AK144680.1 | - | 10 | 1092 | |||
AK146112.1 | - | 10 | 1091 | |||
AK149320.1 | - | 10 | 1092 | |||
BC012638.1 | - | 9 | 950 | 4191809 |