Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfap20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008187.2 | NP_032213.2 | 1, 2 | 380 | ||
GenBank mRNA | AK088713.1 | - | 1, 2 | 284 | ||
AK172603.1 | - | 1, 2 | 356 | |||
BC092397.1 | - | 1, 2 | 268 | |||
BC119023.1 | - | 1, 2 | 276 | 8733772 | ||
BC119025.1 | - | 1, 2 | 276 | 8733774 | ||
BM934726.1 | - | 1, 2 | 380 | |||
Z54179.1 | - | 1, 2 | 303 |