Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trip12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133975.4 | NP_598736.4 | 17 | 2548 | ||
GenBank mRNA | AK041991.1 | - | 1 | 1111 | ||
AK158807.1 | - | 17 | 2548 | |||
BC044869.1 | - | 14 | 1506 |