Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gzmc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010371.2 | NP_034501.2 | 4 | 568 | ||
GenBank mRNA | BC120755.1 | - | 4 | 518 | 40129678 | |
BC120757.1 | - | 4 | 518 | 40129680 | ||
BC145030.1 | - | 4 | 518 | 9053578 | ||
M12301.1 | - | 1 | 22 | |||
M18459.1 | - | 4 | 568 | |||
X12822.1 | - | 4 | 512 |