Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gzme | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010373.3 | NP_034503.2 | 4 | 529 | ||
GenBank mRNA | J03256.1 | - | 4 | 441 | ||
M36901.1 | - | 4 | 421 | |||
X14093.1 | - | 4 | 421 |
Gene Symbol: Gzmd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010372.2 | NP_034502.2 | 4 | 543 | ||
GenBank mRNA | AK131671.1 | - | 4 | 544 | ||
BC141122.1 | - | 4 | 498 | 9055734 | ||
BC145433.1 | - | 498 | 9053991 | |||
BC145434.1 | - | 4 | 498 | |||
J03255.1 | - | 4 | 558 |