Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159551.1 | NP_001153023.1 | 3 | 434 | ||
NM_001159552.1 | NP_001153024.1 | 3 | 434 | |||
NM_001159553.1 | NP_001153025.1 | 3 | 434 | |||
NM_010376.4 | NP_034506.1 | 3 | 434 | |||
GenBank mRNA | AF483214.1 | - | 3 | 401 | ||
AF515662.1 | - | 3 | 313 | |||
AJ345032.1 | - | 3 | 444 | |||
AK004690.1 | - | 3 | 408 | |||
AK007664.1 | - | 3 | 416 | |||
AK012600.1 | - | 3 | 427 | |||
AK019877.1 | - | 3 | 408 | |||
AK028109.1 | - | 3 | 403 | |||
AK039008.1 | - | 3 | 408 | |||
AK049101.1 | - | 3 | 408 | |||
AK050664.1 | - | 3 | 377 | |||
AK051151.1 | - | 3 | 386 | |||
AK080966.1 | - | 3 | 396 | |||
AK159176.1 | - | 3 | 408 | |||
AK169922.1 | - | 3 | 391 | |||
AK171790.1 | - | 3 | 362 | |||
BC056977.1 | - | 3 | 433 | 6818912 | ||
CA321999.1 | - | 3 | 434 | |||
DQ168449.1 | - | 3 | 450 |