Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077709.1 | NP_001071177.1 | 5 | 896 | ||
NM_008202.2 | NP_032228.2 | 4 | 1181 | |||
GenBank mRNA | AK087839.1 | - | 4 | 876 | ||
AK145956.1 | - | 3 | 1146 | |||
AK159968.1 | - | 4 | 1057 | |||
AK167344.1 | - | 3 | 1229 | |||
BC115426.2 | - | 4 | 1035 | 40093613 | ||
BC115427.1 | - | 4 | 1035 | 40093614 | ||
KJ650218.1 | - | 4 | 749 | |||
M32010.1 | - | 4 | 1049 |