Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077709.1 | NP_001071177.1 | 3, 4 | 748 | ||
NM_008202.2 | NP_032228.2 | 2, 3 | 1033 | |||
GenBank mRNA | AK087839.1 | - | 2, 3 | 728 | ||
AK145956.1 | - | 1, 2 | 998 | |||
AK159968.1 | - | 2, 3 | 909 | |||
AK167344.1 | - | 1, 2 | 1081 | |||
BC115426.2 | - | 2, 3 | 887 | 40093613 | ||
BC115427.1 | - | 2, 3 | 887 | 40093614 | ||
KJ650218.1 | - | 2, 3 | 601 | |||
M32010.1 | - | 2, 3 | 901 |