Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2-Ob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010389.3 | NP_034519.2 | 2 | 201 | ||
GenBank mRNA | AK053233.1 | - | 2 | 250 | ||
AK155144.1 | - | 2 | 151 | |||
AK155650.1 | - | 2 | 201 | |||
AK172191.1 | - | 2 | 206 | |||
AK172273.1 | - | 2 | 176 | |||
AK172299.1 | - | 2 | 176 | |||
BC100707.1 | - | 2 | 130 | |||
BC100709.1 | - | 2 | 127 | |||
BC100710.1 | - | 2 | 129 | |||
BC127027.1 | - | 2 | 134 | 40044586 | ||
BC127028.1 | - | 2 | 134 | 40044587 | ||
BY205160.1 | - | 2 | 202 | |||
KJ650230.1 | - | 2 | 134 | |||
KJ663722.1 | - | 2 | 134 |