Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2-Ob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010389.3 | NP_034519.2 | 3 | 446 | ||
GenBank mRNA | AK053233.1 | - | 3 | 495 |