Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2-T18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK153910.1 | - | 5 | 926 |
Gene Symbol: H2-T3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC040514.1 | - | 998 | |||
X03052.1 | - | 572 |
Gene Symbol: H2-T3-like | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY989873.1 | - | 4 | 1137 |