Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2-T3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008208.4 | NP_032234.3 | 5, 6 | 1035 | ||
GenBank mRNA | AK033602.1 | - | 5, 6 | 1036 | ||
BC125378.1 | - | 5, 6 | 1033 | 40129893 | ||
BC125380.1 | - | 5, 6 | 1033 | 40129895 | ||
D86082.1 | - | 5, 6 | 1012 | |||
X03051.1 | - | 4, 5 | 924 |
Gene Symbol: Gm38417 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK169750.1 | - | 1222 |
Gene Symbol: H2-T3-like | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY989871.1 | - | 5, 6 | 1006 |